Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PI 3-Kδ Inhibitor X, IC87114

Catalog No. 5281185MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-811-85MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-811-85MG Supplier MilliporeSigma™ Supplier No. 5281185MG
Add to Cart
Add to Cart

The PI 3-Kδ Inhibitor X, IC87114, also referenced under CAS 371242-69-2, controls the biological activity of PI 3-Kδ. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

A cell permeable, selective inhibitor of PI 3-Kδ (IC50= 1.82, 0.07, and 1.24 for p110b, p110d, and p110g, respectively, but may also target p110a at concentrations as high as 100μM with less potency) in NB-4 and primary APL cells. It has been reported that the constitutive activation of PI3K/Akt signaling in primary AML cells is predominantly attributable to PI 3-Kδ. Pharmacologic use of PI 3-Kδ inhibitor has been shown to block PI3K/Akt/mTOR signaling and could thereby reduce proliferation and induce apoptosis. A related study further suggests PI 3-Kδ inhibitor as a key therapeutic target among PI 3-K isoforms for allergy- and mast cell-related diseases.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
CAS 371242-69-2
Molecular Formula C22H19N7O
Formula Weight 397.44g/mol
Solubility DMSO
Form Powder
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.