accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37131-87-6

IUPAC Name: 5-bromopyrimidine-2-carboxylic acid
Synonyms: 5-bromopyrimidine-2-carboxylic acid
Molecular Weight (g/mol): 203
Molecular Formula: C5H3BrN2O2
InChi Key: XGPTXUYKEDPXCO-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC=C(Br)C=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results