accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37159-60-7

IUPAC Name: 3-bromo-5-chloro-1,2,4-thiadiazole
Synonyms: 3-bromo-5-chloro-1,2,4-thiadiazole
Molecular Weight (g/mol): 199.45
Molecular Formula: C2BrClN2S
InChi Key: CXUWGEWQRCXJDC-UHFFFAOYSA-N
SMILES: ClC1=NC(Br)=NS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results