Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-chloro-1,2,4-thiadiazole, 97%

Catalog No. AC297910100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC297910100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC297910100 Supplier Thermo Scientific Chemicals Supplier No. 297910100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 37159-60-7 | C2BrClN2S | 199.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37159-60-7
Molecular Formula C2BrClN2S
Molecular Weight (g/mol) 199.45
MDL Number MFCD00799287
InChI Key CXUWGEWQRCXJDC-UHFFFAOYSA-N
Synonym 3-bromo-5-chloro-1,2,4 thiadiazole, 1,2,4-thiadiazole, 3-bromo-5-chloro, 3-bromo-5-chloro-1,2,4 thiadiazol, 1,2,4-thiadiazole,3-bromo-5-chloro
PubChem CID 2725053
IUPAC Name 3-bromo-5-chloro-1,2,4-thiadiazole
SMILES ClC1=NC(Br)=NS1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 24.0°C to 25.0°C
Color Colorless to Yellow
Boiling Point 192.0°C to 194.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5965 to 1.5985 (20°C, 589nm)
Quantity 10 g
Solubility Information Solubility in water: slightly soluble in water
Formula Weight 199.46
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Bromo-5-chloro-1, 2, 4-thiadiazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.