accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3717-29-1

IUPAC Name: N-[(3-nitrophenyl)methylidene]hydroxylamine
Synonyms: N-[(3-nitrophenyl)methylidene]hydroxylamine
Molecular Weight (g/mol): 166.14
Molecular Formula: C7H6N2O3
InChi Key: GQMMRLBWXCGBEV-UHFFFAOYSA-N
SMILES: ON=CC1=CC(=CC=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results