Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

syn-3-Nitrobenzaldoxime 98.0+%, TCI America™
SDP

Catalog No. N013225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N013225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N013225G Supplier TCI America Supplier No. N013225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3717-29-1
Molecular Formula C7H6N2O3
Molecular Weight (g/mol) 166.14
MDL Number MFCD00007275
InChI Key GQMMRLBWXCGBEV-UHFFFAOYSA-N
PubChem CID 5369356
IUPAC Name N-[(3-nitrophenyl)methylidene]hydroxylamine
SMILES ON=CC1=CC(=CC=C1)[N+]([O-])=O

Specifications

Melting Point 121°C
Color White-Yellow
Quantity 25 g
Formula Weight 166.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material syn-3-Nitrobenzaldoxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.