accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 371770-32-0

OOHOONH(S)

CAS RN 371770-32-0

IUPAC Name: (2S)-3-cyclopentyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2S)-3-cyclopentyl-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 379.46
Molecular Formula: C23H25NO4
InChi Key: NVZVRXJTMCMDNR-NRFANRHFSA-N
SMILES: OC(=O)[C@H](CC1CCCC1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)