CAS RN 371942-69-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ PIKfyve Inhibitor
The PIKfyve Inhibitor, also referenced under CAS 371942-69-7, controls the biological activity of PIKfyve. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.