accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 372-46-3

IUPAC Name: fluorocyclohexane
Synonyms: cyclohexane, fluoro-
Molecular Weight (g/mol): 102.15
Molecular Formula: C6H11F
InChi Key: GOBGVVAHHOUMDK-UHFFFAOYSA-N
SMILES: FC1CCCCC1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)