Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorocyclohexane, 97%

Catalog No. AC459320250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459320250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC459320250 Supplier Thermo Scientific Chemicals Supplier No. 459320250

missing translation for 'validMainframeMsg'

CAS: 372-46-3 | C6H11F | 102.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-46-3
Molecular Formula C6H11F
Molecular Weight (g/mol) 102.15
InChI Key GOBGVVAHHOUMDK-UHFFFAOYSA-N
Synonym cyclohexane, fluoro, cyclohexyl fluoride, cyclohexylfluoride, fluoro-cyclohexane, acmc-1cpcv
PubChem CID 78988
IUPAC Name fluorocyclohexane
SMILES C1CCC(CC1)F

Specifications

Melting Point 13.0°C
Density 0.9280g/mL
Boiling Point 103.0°C to 105.0°C
Flash Point 5°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula C6H11F
Quantity 25 g
Specific Gravity 0.928
Formula Weight 102.15
Percent Purity 97%
Chemical Name or Material Fluorocyclohexane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
EINECSNumber 206-754-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.