accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 372521-91-0

IUPAC Name: (4-phenylnaphthalen-1-yl)boronic acid
Synonyms: 4-phenylnaphthalen-1-ylboronic acid
Molecular Weight (g/mol): 248.09
Molecular Formula: C16H13BO2
InChi Key: BSKLSKWOKGVQHF-UHFFFAOYSA-N
SMILES: OB(O)C1=C2C=CC=CC2=C(C=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results