Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenylnaphthalene-1-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. P22901G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P22901G Supplier TCI America Supplier No. P22901G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 372521-91-0
Molecular Formula C16H13BO2
Molecular Weight (g/mol) 248.088
MDL Number MFCD16294428
InChI Key BSKLSKWOKGVQHF-UHFFFAOYSA-N
Synonym 4-phenylnaphthalen-1-yl boronic acid, 4-phenylnaphthalene-1-boronic acid, 4-phenyl naphthalene-1-yl boronic acid, 1-phenylnaphthalene-4-boronic acid, 4-phenylnaphthalen-1-yl boronicacid, 4-phenyl naphthalene-1-yl boronic acid;boronic acid,b-4-henyl-1-aphthalenyl
PubChem CID 45382260
IUPAC Name (4-phenylnaphthalen-1-yl)boronic acid
SMILES B(C1=CC=C(C2=CC=CC=C12)C3=CC=CC=C3)(O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 248.09
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylnaphthalene-1-boronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.