accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3C

CAS RN 3726-47-4

IUPAC Name: 1-ethyl-3-methylcyclopentane
Synonyms: 1-methyl-3-ethylcyclopentane
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: PQXAPVOKLYINEI-UHFFFAOYNA-N
SMILES: CCC1CCC(C)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results