CAS RN 37264-66-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Supelco™ Amberlite™ IRA910 Chloride Form
Strongly basic (type II) anion exchange resin for water conditioning and deionization, removal of sulfuric acid and color, etc.