accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3731-39-3

IUPAC Name: N,N-dimethyl-4-[2-(2-methylphenyl)diazen-1-yl]aniline
Synonyms: N,N-dimethyl-4-[2-(2-methylphenyl)diazen-1-yl]aniline
Molecular Weight (g/mol): 239.32
Molecular Formula: C15H17N3
InChi Key: PREOKNMPMCPQQJ-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results