Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)-2'-methylazobenzene, TCI America™
SDP

Catalog No. M07891G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M07891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M07891G Supplier TCI America Supplier No. M07891G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3731-39-3
Molecular Formula C15H17N3
Molecular Weight (g/mol) 239.32
MDL Number MFCD00025648
InChI Key PREOKNMPMCPQQJ-UHFFFAOYSA-N
Synonym 2′C-Methyl-4-(dimethylamino)azobenzene
PubChem CID 19508
IUPAC Name N,N-dimethyl-4-[2-(2-methylphenyl)diazen-1-yl]aniline
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C

Specifications

Melting Point 74°C
Quantity 1 g
Formula Weight 239.32
Physical Form Crystalline Powder
Chemical Name or Material 4-(Dimethylamino)-2′-methylazobenzene
RTECSNumber BX8300000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.