CAS RN 373432-09-7
CAS RN 373432-09-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich [1,3-Bis(Cyclohexyl)Imidazol-2-Ylidene][1,3-Divinyl-1,1,3,3-Tetramethyldisiloxane]Platinum(0)
Sigma Aldrich [1,3-Bis(Cyclohexyl)Imidazol-2-Ylidene][1,3-Divinyl-1,1,3,3-Tetramethyldisiloxane]Platinum(0)
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.