accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3COHOHO

CAS RN 3739-30-8

IUPAC Name: 2-hydroxy-2-methylbutanoic acid
Synonyms: 2-hydroxy-2-methylbutyric acid
Molecular Weight (g/mol): 118.13
Molecular Formula: C5H10O3
InChi Key: MBIQENSCDNJOIY-UHFFFAOYNA-N
SMILES: CCC(C)(O)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Physical Form 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (4)