Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-2-methylbutyric Acid, 95+%, Spectrum™ Chemical
SDP

Catalog No. 18608222 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
25 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-222 25 kg Poly Bottle
18-608-219 5 g Poly Bottle
18-608-220 25 g Poly Bottle
18-608-221 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-608-222 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H403625KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3739-30-8
Molecular Weight (g/mol) 118.13 g/mol

Specifications

CAS Max % 1
Color White
Boiling Point 237.6°C
Melting Range 72°C to 75°C
Packaging Poly Bottle
Quantity 25 kg
Percent Purity 95%
Grade Reagent
Physical Form Crystalline Powder or Lumps
Chemical Name or Material 2-Hydroxy-2-methylbutyric Acid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.