accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3740-92-9

ClClNN

CAS RN 3740-92-9

IUPAC Name: 4,6-dichloro-2-phenylpyrimidine
Synonyms: fenclorim
Molecular Weight (g/mol): 225.07
Molecular Formula: C10H6Cl2N2
InChi Key: NRFQZTCQAYEXEE-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=NC(=N1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results