accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSOO

CAS RN 3741-38-6

IUPAC Name: 1,3,2λ⁴-dioxathiolan-2-one
Synonyms: 1,3,2-dioxathiolane, 2-oxide
Molecular Weight (g/mol): 108.11
Molecular Formula: C2H4O3S
InChi Key: WDXYVJKNSMILOQ-UHFFFAOYSA-N
SMILES: O=S1OCCO1
Boiling Point 
  • (2)
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)