Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethylene sulfite

Catalog No. 77425125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-425-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 77-425-125G Supplier Sigma Aldrich Supplier No. 77425125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Ethylene sulfite is used as a film forming and high temperature additive for electrolytes in lithium ion batteries. It improves decomposition resistance of electrolyte.

Specifications

CAS 3741-38-6
Density 1.426 g/mL at 25°C; 1.433 g/mL (at 25°C)
Boiling Point 159.1°C
Molecular Formula C2H4O3S
Refractive Index n20/D 1.445 (literature); n20/D 1.447
Linear Formula C2H4O3S
Quantity 25 g
MDL Number MFCD00005354
Synonym 1,3,2-Dioxathiolan-2-oxide; Cyclic ethylene sulfite; ES; Glycol sulfite
Molecular Weight (g/mol) 108.12
Percent Purity ≥99.0%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.