accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHN(R)OO

CAS RN 37440-01-0

IUPAC Name: (2R)-2-acetamido-3-(naphthalen-2-yl)propanoate
Synonyms: (2R)-2-acetamido-3-(naphthalen-2-yl)propanoate
Molecular Weight (g/mol): 256.28
Molecular Formula: C15H14NO3
InChi Key: HGTIILKZSFKZMS-CQSZACIVSA-M
SMILES: CC(=O)N[C@H](CC1=CC=C2C=CC=CC2=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)