accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3

CAS RN 374538-05-3

IUPAC Name: 3,8-dimethoxynaphthalene-2-carbaldehyde
Synonyms: 3,8-dimethoxynaphthalene-2-carbaldehyde
Molecular Weight (g/mol): 216.24
Molecular Formula: C13H12O3
InChi Key: YBCQBSORAFCHGW-UHFFFAOYSA-N
SMILES: COC1=CC2=CC=CC(OC)=C2C=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results