accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NFBr

CAS RN 374633-36-0

IUPAC Name: 2-bromo-3-fluoro-6-methylpyridine
Synonyms: 2-bromo-3-fluoro-6-methylpyridine
Molecular Weight (g/mol): 190.02
Molecular Formula: C6H5BrFN
InChi Key: KKVUMKHUMBXLMH-UHFFFAOYSA-N
SMILES: CC1=NC(Br)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results