accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 375-16-6

IUPAC Name: heptafluorobutanoyl chloride
Synonyms: heptafluorobutanoyl chloride
Molecular Weight (g/mol): 232.48
Molecular Formula: C4ClF7O
InChi Key: WFELVFKXQJYPSL-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(Cl)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results