Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Perfluorobutyryl chloride

Catalog No. 2579235G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-792-35G 5 g
25-792-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-792-35G Supplier Sigma Aldrich Supplier No. 2579235G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Perfluorobutyryl chloride has been used in the preparation of:
  • Ethylcellulose perfluorobutyrate, non-thrombogenic novel fluoropolymer for gas-exchange membranes
  • 2-perfluoroalkylbenzothiazole

Specifications

Boiling Point 39°C (lit.)
CAS 375-16-6
Quantity 5 g
Molecular Formula C4ClF7O
Linear Formula CF3CF2CF2COCl
MDL Number MFCD00000707
Synonym Heptafluorobutyryl chloride
Molecular Weight (g/mol) 232.48
Percent Purity 98%
Refractive Index n20/D 1.3 (literature)
Density 1.556 g/mL (at 25°C (literature))
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.