accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHSOOFFFFFFFFF

CAS RN 375-73-5

IUPAC Name: nonafluorobutane-1-sulfonic acid
Synonyms: perfluorobutanesulfonic acid
Molecular Weight (g/mol): 300.09
Molecular Formula: C4HF9O3S
InChi Key: JGTNAGYHADQMCM-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)