Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Nonafluorobutane-1-sulfonic acid

Catalog No. 5626295G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-262-95G 5 g
56-262-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 56-262-95G Supplier Sigma Aldrich Supplier No. 5626295G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Nonafluorobutane-1-sulfonic acid can be considered as a superacid, which can be synthesized by reacting 1-iodononafluorobutane with sodium dithionite in the presence of sodium bicarbonate in acetonitrile/water.Nonafluorobutane-1-sulfonic acid may be used as catalyst in the synthesis of 6-chloro-6H-dibenz[c,e][1,2]oxaphosphorin and N-benzylpyridin-2-amine.

Specifications

CAS 375-73-5
Density 1.811 g/mL (at 25°C (literature))
Boiling Point 112°C to 114°C (14 mmHg)
Molecular Formula C4HF9O3S
Refractive Index n20/D 1.3230 (literature)
Linear Formula CF3(CF2)3 SO3H
Quantity 5 g
MDL Number MFCD01320794
Molecular Weight (g/mol) 300.1
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.