accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 375-80-4

IUPAC Name: 1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodohexane
Synonyms: 1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodohexane
Molecular Weight (g/mol): 553.86
Molecular Formula: C6F12I2
InChi Key: JOQDDLBOAIKFQX-UHFFFAOYSA-N
SMILES: FC(F)(I)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results