Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecafluoro-1,6-diiodohexane 98.0+%, TCI America™
SDP

Catalog No. D233310G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D233310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D233310G Supplier TCI America Supplier No. D233310G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 375-80-4
Molecular Formula C6F12I2
Molecular Weight (g/mol) 553.86
MDL Number MFCD00042264
InChI Key JOQDDLBOAIKFQX-UHFFFAOYSA-N
Synonym 1,6-diiodoperfluorohexane, 1,6-diiodododecafluorohexane, dodecafluoro-1,6-diiodohexane, 1,6-diiododecafluorohexane, perfluoro-1,6-diiodohexane, hexane, 1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodo, acmc-209iu0, 1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodo-hexane, hexane,1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodo
PubChem CID 78994
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodohexane
SMILES FC(F)(I)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I

Specifications

Color Red-White
Boiling Point 115°C
Quantity 10 g
Formula Weight 553.86
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Dodecafluoro-1,6-diiodohexane
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.