accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3751-48-2

IUPAC Name: 3-(3-methylphenyl)propanoic acid
Synonyms: 3-(3-methylphenyl)propanoic acid
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: VWXVTHDQAOAENP-UHFFFAOYSA-N
SMILES: CC1=CC=CC(CCC(O)=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results