accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3752-25-8

IUPAC Name: (2E)-3-(2-chlorophenyl)prop-2-enoic acid
Synonyms: 2-chlorocinnamic acid
Molecular Weight (g/mol): 182.6
Molecular Formula: C9H7ClO2
InChi Key: KJRRTHHNKJBVBO-AATRIKPKSA-N
SMILES: OC(=O)\C=C\C1=CC=CC=C1Cl
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)