Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorocinnamic acid, predominantly trans, 99%, Thermo Scientific™

Catalog No. AAL0160622
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0160622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0160622 Supplier Thermo Scientific Chemicals Supplier No. L0160622

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3752-25-8
Molecular Formula C9H7ClO2
Molecular Weight (g/mol) 182.603
MDL Number MFCD00004372
InChI Key KJRRTHHNKJBVBO-AATRIKPKSA-N
Synonym 2-chlorocinnamic acid, o-chlorocinnamic acid, 3-2-chlorophenyl acrylic acid, 2e-3-2-chlorophenyl acrylic acid, e-3-2-chlorophenyl acrylic acid, cinnamic acid, o-chloro, 2e-3-2-chlorophenyl prop-2-enoic acid, 2-propenoic acid, 3-2-chlorophenyl, e-o-chlorocinnamic acid, 3-2-chlorophenyl-2-propenoic acid
PubChem CID 700642
IUPAC Name (E)-3-(2-chlorophenyl)prop-2-enoic acid
SMILES C1=CC=C(C(=C1)C=CC(=O)O)Cl

Specifications

Melting Point 209°C to 212°C
Quantity 100 g
Beilstein 1365198
Formula Weight 182.61
Percent Purity 99%
Chemical Name or Material 2-Chlorocinnamic acid, predominantly trans
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECSNumber GD8450000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.