accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37526-88-8

IUPAC Name: benzyl (2E)-2-methylbut-2-enoate
Synonyms: benzyl tiglate
Molecular Weight (g/mol): 190.24
Molecular Formula: C12H14O2
InChi Key: QRGSTISKDZCDHV-XCVCLJGOSA-N
SMILES: C\C=C(/C)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results