Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Tiglate 97.0+%, TCI America™
SDP

Catalog No. B334825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B334825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B334825G Supplier TCI America Supplier No. B334825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 37526-88-8
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.24
MDL Number MFCD00017279
InChI Key QRGSTISKDZCDHV-XCVCLJGOSA-N
Synonym benzyl tiglate, benzyl 2-methylcrotonate, benzyl trans-2,3-dimethylacrylate, e-benzyl 2-methylbut-2-enoate, benzyl trans-2-methyl-2-butenoate, unii-67uep899rr, benzyl 2-methyl-2-butenoate, benzyl trans-2-methylcrotonate, tiglic acid benzyl ester, benzyltiglate
PubChem CID 250096
IUPAC Name benzyl (2E)-2-methylbut-2-enoate
SMILES C\C=C(/C)C(=O)OCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 134°C
Quantity 25 g
Formula Weight 190.24
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Tiglate
EINECSNumber (3)-1047
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.