accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3755-83-7

CH3SN

CAS RN 3755-83-7

IUPAC Name: 2-methyl-4,5-diphenyl-1,3-thiazole
Synonyms: 2-methyl-4,5-diphenyl-1,3-thiazole
Molecular Weight (g/mol): 251.35
Molecular Formula: C16H13NS
InChi Key: ZFYWCVZNRMSXBT-UHFFFAOYSA-N
SMILES: CC1=NC(=C(S1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results