accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHIN

CAS RN 375853-79-5

IUPAC Name: tert-butyl N-(5-iodopyridin-2-yl)carbamate
Synonyms: tert-butyl N-(5-iodopyridin-2-yl)carbamate
Molecular Weight (g/mol): 320.13
Molecular Formula: C10H13IN2O2
InChi Key: YZLXIVKPYUFJPN-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC=C(I)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results