accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONBr

CAS RN 375857-64-0

IUPAC Name: 5-(2-bromophenyl)-3-methyl-1,2,4-oxadiazole
Synonyms: 5-(2-bromophenyl)-3-methyl-1,2,4-oxadiazole
Molecular Weight (g/mol): 239.07
Molecular Formula: C9H7BrN2O
InChi Key: NYAFKUCVAVUCHW-UHFFFAOYSA-N
SMILES: CC1=NOC(=N1)C1=CC=CC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results