accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37609-25-9

IUPAC Name: (5Z)-cyclohexadec-5-en-1-one
Synonyms: 5-cyclohexadecenone
Molecular Weight (g/mol): 236.4
Molecular Formula: C16H28O
InChi Key: ABRIMXGLNHCLIP-VURMDHGXSA-N
SMILES: O=C1CCCCCCCCCC\C=C/CCC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results