CAS RN 3761-53-3
Xylidine Ponceau 2R, pure
CAS: 3761-53-3 Molecular Formula: C18H14N2Na2O7S2 Molecular Weight (g/mol): 480.417 MDL Number: MFCD00003897 InChI Key: YJVBLROMQZEFPA-UHFFFAOYSA-L Synonym: 4-(2,4-Dimethylphenylazo)-3-hydroxy-2,7-naphthalenedisulfonic acid, disodium salt,C.I. 16150,4-(2, 4-Dimethylphenylazo)-3-hydroxy-2, 7-naphthalenedisulfonic acid,Ponceau G, R, 2R PubChem CID: 102092631 ChEBI: CHEBI:82369 IUPAC Name: disodium;5-[(2,4-dimethylphenyl)diazenyl]-6-oxido-7-sulfonaphthalene-2-sulfonate SMILES: CC1=CC(=C(C=C1)N=NC2=C3C=CC(=CC3=CC(=C2[O-])S(=O)(=O)O)S(=O)(=O)[O-])C.[Na+].[Na+]
Acid Red 26, TCI America™
CAS: 3761-53-3 Molecular Formula: C18H14N2Na2O7S2 Molecular Weight (g/mol): 480.417 MDL Number: MFCD00003897 InChI Key: YJVBLROMQZEFPA-UHFFFAOYSA-L Synonym: Ponceau de Xylidine, Ponceau R PubChem CID: 102092631 ChEBI: CHEBI:82369 IUPAC Name: disodium;5-[(2,4-dimethylphenyl)diazenyl]-6-oxido-7-sulfonaphthalene-2-sulfonate SMILES: CC1=CC(=C(C=C1)N=NC2=C3C=CC(=CC3=CC(=C2[O-])S(=O)(=O)O)S(=O)(=O)[O-])C.[Na+].[Na+]
MP Biomedicals, Inc Ponceau 2R MP Biomedicals
CAS: 3761-53-3 Molecular Formula: C18H14N2Na2O7S2 Molecular Weight (g/mol): 480.417 InChI Key: YJVBLROMQZEFPA-UHFFFAOYSA-L Synonym: Xylidene Ponceau 2R PubChem CID: 102092631 ChEBI: CHEBI:82369 IUPAC Name: disodium;5-[(2,4-dimethylphenyl)diazenyl]-6-oxido-7-sulfonaphthalene-2-sulfonate SMILES: CC1=CC(=C(C=C1)N=NC2=C3C=CC(=CC3=CC(=C2[O-])S(=O)(=O)O)S(=O)(=O)[O-])C.[Na+].[Na+]
Brilliant Ponceau G, MP Biomedicals
CAS: 3761-53-3 Molecular Formula: C18H14N2Na2O7S2 Molecular Weight (g/mol): 480.417 InChI Key: YJVBLROMQZEFPA-UHFFFAOYSA-L PubChem CID: 102092631 ChEBI: CHEBI:82369 IUPAC Name: disodium;5-[(2,4-dimethylphenyl)diazenyl]-6-oxido-7-sulfonaphthalene-2-sulfonate SMILES: CC1=CC(=C(C=C1)N=NC2=C3C=CC(=CC3=CC(=C2[O-])S(=O)(=O)O)S(=O)(=O)[O-])C.[Na+].[Na+]