Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brilliant Ponceau G, MP Biomedicals

Catalog No. ICN214339 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN214339 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN214339 Supplier MP Biomedicals, Inc Supplier No. 0521433901

Chemical Identifiers

CAS 3761-53-3
Molecular Formula C18H14N2Na2O7S2
Molecular Weight (g/mol) 480.417
InChI Key YJVBLROMQZEFPA-UHFFFAOYSA-L
PubChem CID 102092631
ChEBI CHEBI:82369
IUPAC Name disodium;5-[(2,4-dimethylphenyl)diazenyl]-6-oxido-7-sulfonaphthalene-2-sulfonate
SMILES CC1=CC(=C(C=C1)N=NC2=C3C=CC(=CC3=CC(=C2[O-])S(=O)(=O)O)S(=O)(=O)[O-])C.[Na+].[Na+]

Specifications

Formula Weight 480.4
Quantity 10 g
Chemical Name or Material Brilliant Ponceau G
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.