accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CMgBr

CAS RN 3761-92-0

IUPAC Name: bromo(hexyl)magnesium
Synonyms: bromo(hexyl)magnesium
Molecular Weight (g/mol): 189.38
Molecular Formula: C6H13BrMg
InChi Key: SFXUWNMDTWQSHE-UHFFFAOYSA-M
SMILES: CCCCCC[Mg]Br
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)