Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexylmagnesium bromide, 0.8M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog No. AC427768000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427768000 800 mL AcroSeal™ Glass Bottle
AC427761000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC427768000 Supplier Thermo Scientific Chemicals Supplier No. 427768000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3761-92-0 | Molecular Formula: C6H13BrMg | Molecular Weight: 189.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3761-92-0 , 109-99-9
Molecular Formula C6H13BrMg
Molecular Weight (g/mol) 189.38
MDL Number MFCD00000046
InChI Key SFXUWNMDTWQSHE-UHFFFAOYSA-M
Synonym hexylmagnesium bromide, magnesium, bromohexyl, n-hexylmagnesium bromide, n-hexmgbr, hexyl magnesium bromide, n-hexyl magnesium bromide, hexylmagnesium bromide solution, sfxuwnmdtwqshe-uhfffaoysa-m, hexylmagnesium bromide solution, 2.0 m in diethyl ether, grignard reagent
PubChem CID 4184939
IUPAC Name magnesium;hexane;bromide
SMILES CCCCCC[Mg]Br

Specifications

Color Brown
Density 0.9800g/mL
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3(CH2)5MgBr
Quantity 800 mL
Specific Gravity 0.98
Formula Weight 189.39
Physical Form Liquid
Chemical Name or Material Hexylmagnesium bromide, 0.8M solution in THF
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.