accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3CH3

CAS RN 37615-53-5

IUPAC Name: dihexyl(methyl)amine
Synonyms: dihexyl(methyl)amine
Molecular Weight (g/mol): 199.38
Molecular Formula: C13H29N
InChi Key: POMGZMHIXYRARC-UHFFFAOYSA-N
SMILES: CCCCCCN(C)CCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results