Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Di-n-hexylmethylamine, MP Biomedicals

Catalog No. ICN227585 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN227585 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN227585 Supplier MP Biomedicals, Inc Supplier No. 0522758525

Chemical Identifiers

CAS 37615-53-5
Molecular Formula C13H29N
Molecular Weight (g/mol) 199.382
InChI Key POMGZMHIXYRARC-UHFFFAOYSA-N
Synonym n-methyldihexylamine, methyldihexylamine, n,n-di-n-hexyl methylamine, dihexyl methyl amine, n,n-dihexylmethylamine, acmc-1agc7, n,n-di-n-hexylmethylamine
PubChem CID 4426313
IUPAC Name N-hexyl-N-methylhexan-1-amine
SMILES CCCCCCN(C)CCCCCC

Specifications

Quantity 100 g
Formula Weight 199.37
Chemical Name or Material N, N-Di-n-Hexylmethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.