accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHOHNO

CAS RN 376382-11-5

IUPAC Name: 16-[(2-phenylethyl)amino]-14-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1(16),2,4,6,9(17),10,12-heptaene-8,15-dione
Synonyms: 16-[(2-phenylethyl)amino]-14-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1(16),2,4,6,9(17),10,12-heptaene-8,15-dione
Molecular Weight (g/mol): 366.42
Molecular Formula: C24H18N2O2
InChi Key: XASCINRGTHLHGM-UHFFFAOYSA-N
SMILES: O=C1NC2=CC=CC3=C2C(=C1NCCC1=CC=CC=C1)C1=CC=CC=C1C3=O
Product Line 
  • (2)
Quantity 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results