Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Insulin Expression Inducer, BRD7389

Catalog No. 20370910MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-370-910MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-370-910MG Supplier MilliporeSigma™ Supplier No. 20370910MG
Add to Cart
Add to Cart

A cell-permeable naphthoquinolinedione compound that specifically induces insulin secretion in mouse αTC1-cells and promotes transdifferentiation to a β-cell-like status.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Orange-yellow
Description 98% by HPLC
CAS 376382-11-5
Molecular Formula C24H18N2O2
Formula Weight 366.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.