accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 376609-78-8

IUPAC Name: 4-(octadecyloxy)-1,1'-biphenyl
Synonyms: 4-(octadecyloxy)-1,1'-biphenyl
Molecular Weight (g/mol): 422.7
Molecular Formula: C30H46O
InChi Key: WJPFTOGDAJEWPP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCOC1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results