Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Octadecyloxybiphenyl 98.0+%, TCI America™
SDP

Catalog No. B19245G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19245G 5 g
B192425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19245G Supplier TCI America Supplier No. B19245G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 376609-78-8
Molecular Formula C30H46O
Molecular Weight (g/mol) 422.697
MDL Number MFCD01321145
InChI Key WJPFTOGDAJEWPP-UHFFFAOYSA-N
Synonym Biphenyl-4-yl Octadecyl Ether, Biphenyl-4-yl Stearyl Ether, 4-Stearyloxybiphenyl
PubChem CID 22899691
IUPAC Name 1-octadecoxy-4-phenylbenzene
SMILES CCCCCCCCCCCCCCCCCCOC1=CC=C(C=C1)C2=CC=CC=C2

Specifications

Melting Point 92°C
Color White
Quantity 5 g
Formula Weight 422.70
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Octadecyloxybiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.